Skip to Content
Merck
All Photos(1)

Key Documents

184365

Sigma-Aldrich

(Methylsulfonyl)acetonitrile

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€242.00

€242.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€242.00

About This Item

Linear Formula:
CH3SO2CH2CN
CAS Number:
Molecular Weight:
119.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€242.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

81-84 °C (lit.)

functional group

nitrile
sulfone

SMILES string

CS(=O)(=O)CC#N

InChI

1S/C3H5NO2S/c1-7(5,6)3-2-4/h3H2,1H3

InChI key

FOTRKCAZUSJCQD-UHFFFAOYSA-N

Application

(Methylsulfonyl)acetonitrile was used in the synthesis of:
  • substituted δ-pyrone ring analogues[1]
  • 4,9-dihydropyrrolo[2,1-b]quinazolines containing electron withdrawing groups at the 3-position[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel entry to substituted chromones and furochromones through cyclopropane intermediates.
Gammill RB, et al.
Tetrahedron Letters, 33(8), 997-1000 (1992)
An Intramolecular N-Arylation Approach to 3-Functionalized 4, 9-Dihydropyrrolo [2, 1-b] quinazolines.
Suthiwangcharoen N, et al.
Journal of Heterocyclic Chemistry, 48(3), 706-709 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service