Skip to Content
Merck
All Photos(1)

Key Documents

184241

Sigma-Aldrich

4-Butoxybenzyl alcohol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OC6H4CH2OH
CAS Number:
Molecular Weight:
180.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

solid

refractive index

n20/D 1.518 (lit.)

mp

29-32 °C (lit.)

SMILES string

CCCCOc1ccc(CO)cc1

InChI

1S/C11H16O2/c1-2-3-8-13-11-6-4-10(9-12)5-7-11/h4-7,12H,2-3,8-9H2,1H3

InChI key

MGKNBDBZIKPUFM-UHFFFAOYSA-N

Application

4-Butoxybenzyl alcohol was used in the synthesis of:
  • 3-(4-butoxyphenyl)-1-phenylpropan-1-ol[1]
  • 3-(4-butoxyphenyl)-1-phenylpropan-1-one[1]
  • 4-butoxybenzyl chloride[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dinakar Gnanamgari et al.
Organic & biomolecular chemistry, 6(23), 4442-4445 (2008-11-14)
Primary alcohols can be coupled with secondary benzylic alcohols by an air-stable catalytic system involving terpyridine ruthenium or iridium complexes.
Molecular recognition directed self-assembly of tubular liquid crystalline and crystalline supramolecular architectures from taper shaped (15-crown-5) methyl 3, 4, 5-tris (p-alkyloxybenzyloxy) benzoates and (15-crown-5) methyl 3, 4, 5-tris (p-dodecyloxy) benzoate.
Johansson G, et al.
Journal of the Chemical Society. Perkin Transactions 1, 4, 447-459 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service