Skip to Content
Merck
All Photos(1)

Key Documents

183601

Sigma-Aldrich

2,5-Dichlorobenzyl alcohol

99%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€130.00

€130.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€130.00

About This Item

Linear Formula:
Cl2C6H3CH2OH
CAS Number:
Molecular Weight:
177.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€130.00


Check Cart for Availability

Request a Bulk Order

Assay

99%

form

powder

mp

79-80 °C (lit.)

functional group

chloro
hydroxyl

SMILES string

OCc1cc(Cl)ccc1Cl

InChI

1S/C7H6Cl2O/c8-6-1-2-7(9)5(3-6)4-10/h1-3,10H,4H2

InChI key

LCEIGNVIDJNUGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,5-Dichlorobenzyl alcohol participates in [Ru(acac)2(CH3CN)2]PF6 (acac = acetylacetone) catalyzed oxidation of alcohols to aldehydes or ketones using H5IO6 as oxidant[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mild oxidation of alcohols with periodic acid catalyzed by [Ru(acac)2(CH3CN)2]PF6 in water.
Ganesamoorthy S, et al.
Catalysis Communications, 10(14), 1835-1838 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service