Skip to Content
Merck
All Photos(1)

Key Documents

183180

Sigma-Aldrich

Tetrathiafulvalene

97%

Synonym(s):

Δ2,2′-Bi-1,3-dithiole, TTF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4S4
CAS Number:
Molecular Weight:
204.36
Beilstein:
1617956
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

116-119 °C (lit.)

SMILES string

S1C=CS\C1=C2/SC=CS2

InChI

1S/C6H4S4/c1-2-8-5(7-1)6-9-3-4-10-6/h1-4H

InChI key

FHCPAXDKURNIOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrathiafulvalene (TTF) is an electron-donor which consists of oligomers, dendrimers and polymers which can be used in the formation of redox macromolecules.[1]

Application

TTF may be linked with lithium chloride (LiCl) to form a precipitated layer on the lithium oxide (Li2O2) for the fabrication of high performance Li-O2 batteries.[2] Bio-sourced carbon nanodots can be surface modified by TTF which can be potentially used for electrochemical applications.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laurence Albiges et al.
European journal of cancer (Oxford, England : 1990), 51(17), 2580-2586 (2015-09-09)
Monoclonal antibodies that target the programmed death-1 (PD-1)/programmed death-ligand 1(PD-L1) pathway have shown antitumour activity in metastatic renal cell carcinoma (mRCC) and are currently being developed in first-line (in combination) and in previously treated patients. The efficacy targeted therapy (TT)
Exploring Tetrathiafulvalene-Carbon Nanodot Conjugates in Charge Transfer Reactions.
Ferrer-Ruiz A, et al.
Angewandte Chemie (International Edition in English), 57(4), 1001-1005 (2018)
Surface-nhanced near-infrared fourier transform Raman scattering of tetrathiafulvalene adsorbed on silver powder
Nie, S., & Yu, N. T.
Journal of Raman Spectroscopy, 22(9), 489-495 (1991)
Aldrichimica Acta, 18, 73-73 (1985)
Chemical Society Reviews, 23, 41-41 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service