Skip to Content
Merck
All Photos(1)

Key Documents

181803

Sigma-Aldrich

Poly(2,6-dimethyl-1,4-phenylene oxide)

Mn 20,000

Synonym(s):

2,6-Dimethyl-1,4-phenylene ether homopolymer, 2,6-Dimethylphenol homopolymer, 2,6-Dimethylphenol polymer, Poly(2,6-xylenol)

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€459.00

€459.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€459.00

About This Item

Linear Formula:
[C6H2(CH3)2O]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€459.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

form

powder

mol wt

Mn 20,000
Mw 16,000-24,000 by GPC

density

1.06 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(O)cc(C)c1O

InChI

1S/C8H10O/c1-6-4-3-5-7(2)8(6)9/h3-5,9H,1-2H3

InChI key

NXXYKOUNUYWIHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

A lot specific analysis is included with each unit

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service