Skip to Content
Merck
All Photos(1)

Key Documents

18165

Sigma-Aldrich

(R)-(+)-2-Bromopropionic acid

≥98.0% (sum of enantiomers, GC)

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
€447.00

€447.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
€447.00

About This Item

Empirical Formula (Hill Notation):
C3H5BrO2
CAS Number:
Molecular Weight:
152.97
Beilstein:
1720262
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

€447.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98.0% (sum of enantiomers, GC)

form

liquid

optical activity

[α]20/D +26±2°, neat

refractive index

n20/D 1.475

density

1.692 g/mL at 20 °C (lit.)

functional group

bromo
carboxylic acid

storage temp.

2-8°C

SMILES string

C[C@@H](Br)C(O)=O

InChI

1S/C3H5BrO2/c1-2(4)3(5)6/h2H,1H3,(H,5,6)/t2-/m1/s1

InChI key

MONMFXREYOKQTI-UWTATZPHSA-N

Caution

may discolor to brownish on storage

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eugénie Grigorian et al.
Frontiers in microbiology, 12, 725997-725997 (2021-10-09)
L-2-halocid dehalogenases (L-2-HADs) have been mainly characterized from terrestrial polluted environments. By contrast, knowledge is still scarce about their role in detoxification of predominant halocarbons in marine environments. Here, phylogenetic analyses showed a wide diversity of homologous L-2-HADs, especially among

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service