Skip to Content
Merck
All Photos(1)

Key Documents

17996

Sigma-Aldrich

(±)-Camphoric acid anhydride

≥98.0% (T)

Synonym(s):

(±)-1,8,8-Trimethyl-3-oxabicyclo[3.2.1]octane-2,4-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O3
CAS Number:
Molecular Weight:
182.22
Beilstein:
6582434
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98.0% (T)

form

solid

mp

222-225 °C

storage temp.

2-8°C

SMILES string

CC1(C)[C@H]2CC[C@]1(C)C(=O)OC2=O

InChI

1S/C10H14O3/c1-9(2)6-4-5-10(9,3)8(12)13-7(6)11/h6H,4-5H2,1-3H3/t6-,10+/m0/s1

InChI key

VFZDNKRDYPTSTP-QUBYGPBYSA-N

Application

(±)-Camphoric acid anhydride (1,8,8-Trimethyl-3-oxabicyclo[3.2.1]octane-2,4-dione) may be useful in the synthesis of aliphatic-aromatic copolyesters.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quentin Ruiz et al.
Polymers, 12(1) (2020-01-23)
Epoxy resin plays a key role in composite matrices and DGEBA is the major precursor used. With the aim of favouring the use of bio resources, epoxy resins can be prepared from lignin. In particular, diglycidyl ether of isoeugenol derivatives

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service