Skip to Content
Merck
All Photos(1)

Documents

178683

Sigma-Aldrich

1,2,3,4-Tetrahydro-2-naphthoic acid

98%

Synonym(s):

Tetralin-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O2
CAS Number:
Molecular Weight:
176.21
Beilstein:
2050435
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

mp

93-96 °C (lit.)

SMILES string

OC(=O)C1CCc2ccccc2C1

InChI

1S/C11H12O2/c12-11(13)10-6-5-8-3-1-2-4-9(8)7-10/h1-4,10H,5-7H2,(H,12,13)

InChI key

NTAGXJQHJQUOOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R U Meckenstock et al.
Applied and environmental microbiology, 66(7), 2743-2747 (2000-07-06)
Anaerobic naphthalene degradation by a sulfate-reducing enrichment culture was studied by substrate utilization tests and identification of metabolites by gas chromatography-mass spectrometry. In substrate utilization tests, the culture was able to oxidize naphthalene, 2-methylnaphthalene, 1- and 2-naphthoic acids, phenylacetic acid
Beili Wang et al.
Environmental science & technology, 53(2), 1022-1030 (2018-12-18)
The nontargeted scanning chemical profiling approach has shown great potential to identify unknown pollutants or novel biological markers; however, the structure identification of unknown compounds is a challenge. In this study, a carboxyl-specific derivatization reagent, N-(4-aminomethylphenyl) pyridinium (AMPP), was coupled
Hazel M Girvan et al.
Journal of inorganic biochemistry, 188, 18-28 (2018-08-18)
The CYP152 family of cytochrome P450 enzymes (P450s or CYPs) are bacterial peroxygenases that use hydrogen peroxide to drive hydroxylation and decarboxylation of fatty acid substrates. We have expressed and purified a novel CYP152 family member - CYP152K6 from the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service