Skip to Content
Merck
All Photos(2)

Key Documents

178608

Sigma-Aldrich

2,4-Dimethoxybenzylamine hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CH2NH2·HCl
CAS Number:
Molecular Weight:
203.67
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

188-190 °C (lit.)

SMILES string

Cl.COc1ccc(CN)c(OC)c1

InChI

1S/C9H13NO2.ClH/c1-11-8-4-3-7(6-10)9(5-8)12-2;/h3-5H,6,10H2,1-2H3;1H

InChI key

KJOWSAVFSCSBMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,4-Dimethoxybenzylamine hydrochloride was used in the synthesis of 2-(2,4-dimethoxybenzylamino)-4-phenyl-3-butenonitrile.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Greenwald RB, et al.
The Journal of Organic Chemistry, 41(8), 1469-1470 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service