Skip to Content
Merck
All Photos(2)

Key Documents

176958

Sigma-Aldrich

Propionaldehyde diethyl acetal

97%

Synonym(s):

1,1-Diethoxypropane

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€36.70

€36.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€36.70

About This Item

Linear Formula:
CH3CH2CH(OC2H5)2
CAS Number:
Molecular Weight:
132.20
Beilstein:
1697731
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€36.70


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.389 (lit.)

bp

122.8 °C (lit.)

density

0.815 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

CCOC(CC)OCC

InChI

1S/C7H16O2/c1-4-7(8-5-2)9-6-3/h7H,4-6H2,1-3H3

InChI key

MBNMGGKBGCIEGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Propionaldehyde diethyl acetal was used as a reducing agent in the aerobic epoxidation of alkenes[1].

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Co (thd)2: a superior catalyst for aerobic epoxidation and hydroperoxysilylation of unactivated alkenes: application to the synthesis of spiro-1, 2, 4-trioxanes.
Tetrahedron Letters, 44(44), 8135-8138 (2003)
Rita De Santis et al.
PloS one, 6(6), e21075-e21075 (2011-06-28)
We recently reported that the oxidized avidin, named AvidinOX®, resides for weeks within injected tissues as a consequence of the formation of Schiff's bases between its aldehyde groups and tissue protein amino groups. We also showed, in a mouse pre-clinical
Runxian Li et al.
Metabolites, 10(1) (2019-12-28)
Copper (Cu) is widely used in the swine industry to improve the growth performance of pigs. However, high doses of copper will induce cell damage and toxicity. The aim of this study was to evaluate toxicity, bioavailability, and effects on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service