Skip to Content
Merck

Skip To

175684

Ammonium-d8 sulfate

98 atom % D

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 g
Check Cart for Availability
€ 137,00

About This Item

Linear Formula:
(ND4)2SO4
CAS Number:
Molecular Weight:
140.19
NACRES:
NA.12
PubChem Substance ID:
UNSPSC Code:
12142200
MDL number:
Isotopic purity:
98 atom % D
Mass shift:
M+8
Form:
solid

€ 137,00


Check Cart for Availability

Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


isotopic purity

98 atom % D

Quality Segment

form

solid

technique(s)

NMR: suitable

mp

>280 °C (dec.) (lit.)

mass shift

M+8

SMILES string

[2H]N([2H])[2H].[2H]N([2H])[2H].[2H]OS(=O)(=O)O[2H]

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)/i/hD8

InChI key

BFNBIHQBYMNNAN-KTOHAENGSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
299286440485175838
description

98 atom % D

description

98 atom % 15N

description

98 atom % D

description

98 atom % D

mass shift

M+8

mass shift

M+2

mass shift

M+7

mass shift

M+5

isotopic purity

98 atom % D

isotopic purity

98 atom % 15N

isotopic purity

98 atom % D

isotopic purity

98 atom % D

technique(s)

NMR: suitable

technique(s)

bio NMR: suitable, protein expression: suitable

technique(s)

NMR: suitable

technique(s)

NMR: suitable, bio NMR: suitable

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

solid

form

solid

form

solid

form

solid

mp

>280 °C (dec.) (lit.)

mp

>280 °C (dec.) (lit.)

mp

110-112 °C (dec.) (lit.)

mp

240 °C (dec.) (lit.)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

13814-01-2

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters