Skip to Content
Merck
All Photos(1)

Key Documents

174416

Sigma-Aldrich

3-Hydroxypiperidine hydrochloride

99%

Synonym(s):

3-Piperidinol hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€93.00

€93.00


Estimated to ship on27 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€93.00

About This Item

Empirical Formula (Hill Notation):
C5H11NO · HCl
CAS Number:
Molecular Weight:
137.61
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€93.00


Estimated to ship on27 May 2025


Request a Bulk Order

Assay

99%

form

solid

SMILES string

Cl[H].OC1CCCNC1

InChI

1S/C5H11NO.ClH/c7-5-2-1-3-6-4-5;/h5-7H,1-4H2;1H

InChI key

VLECDMDGMKPUSK-UHFFFAOYSA-N

Application

3-Hydroxypiperidine hydrochloride was used for the on-line analysis and solution phase synthesis of Ugi multicomponent reaction (MCR) products using miniaturised-SYNthesis and Total Analysis System (μSYNTAS)[1].
Reactant for synthesis of:
Istaroxime analogues as Na+, K+ - ATPase inhibitors
Nitric oxide formation inhibitors
Selective CC chemokine receptor-3 antagonists
Oxygenated carbolines
α-lithioallyl amides

Reactant for highly chemoselective reductive amination of carbonyl compounds

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M C Mitchell et al.
The Analyst, 126(1), 24-27 (2001-02-24)
A miniaturised-SYNthesis and Total Analysis System (mu SYNTAS) was used for the solution-phase synthesis and on-line analysis (TOF-MS) of Ugi multicomponent reaction (MCR) products. This approach provides an unusually high degree of control of the MCR and delivers detailed, novel

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service