Skip to Content
Merck
All Photos(2)

Key Documents

173495

Sigma-Aldrich

Geranyl acetate

≥97%

Synonym(s):

trans-3,7-Dimethyl-2,6-octadien-1-yl acetate, trans-3,7-Dimethyl-2,6-octadienyl acetate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€41.50
100 G
€89.10

€41.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€41.50
100 G
€89.10

About This Item

Linear Formula:
CH3CO2CH2CH=C(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
196.29
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€41.50


Check Cart for Availability

Request a Bulk Order

vapor density

6.8 (vs air)

Quality Level

vapor pressure

0.07 mmHg ( 20 °C)

Assay

≥97%

form

liquid

impurities

1-2% neryl acetate

refractive index

n20/D 1.461 (lit.)

bp

138 °C/25 mmHg (lit.)

density

0.916 g/mL at 25 °C

functional group

ester

storage temp.

2-8°C

SMILES string

CC(=O)OC\C=C(/C)CC\C=C(/C)C

InChI

1S/C12H20O2/c1-10(2)6-5-7-11(3)8-9-14-12(4)13/h6,8H,5,7,9H2,1-4H3/b11-8+

InChI key

HIGQPQRQIQDZMP-DHZHZOJOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Geranyl acetate is an aroma active rice volatile[1]. It is one of the components of the essential oils of Daucus carota L. seeds[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuai Liu et al.
Insect biochemistry and molecular biology, 127, 103485-103485 (2020-10-14)
The sensory neuron membrane protein, SNMP1, was initially discovered in moths and is associated with sex pheromone sensitive neurons, suggesting a role in the detection of these semiochemicals. Although DrosophilaSNMP1 has been reported to be involved in detecting of the
Nesrine Rokbeni et al.
Chemistry & biodiversity, 10(12), 2278-2290 (2013-12-12)
The essential oils of Daucus carota L. (Apiaceae) seeds sampled from ten wild populations spread over northern Tunisia were characterized by GC-FID and GC/MS analyses. In total, 36 compounds were identified in the D. carota seed essential oils, with a
Bruno Pandelo Brügger et al.
Scientific reports, 9(1), 8358-8358 (2019-06-09)
Podisus nigrispinus Dallas (Heteroptera: Pentatomidae), released in biological control programs, is a predator of Lepidopteran and Coleopteran species. Lemongrass essential oil and its constituents can be toxic to this natural enemy. The major constituents of lemongrass essential oil are neral
Satoshi Nojima et al.
PloS one, 6(3), e18178-e18178 (2011-04-06)
Semiochemicals are often produced in infinitesimally small quantities, so their isolation requires large amounts of starting material, not only requiring significant effort in sample preparation, but also resulting in a complex mixture of compounds from which the bioactive compound needs
Nehad E M Taktak et al.
Pesticide biochemistry and physiology, 158, 156-165 (2019-08-06)
Culex pipiens is a main vector for Bancroftian filariasis, Rift Valley Fever and diseases caused by other viruses, leaving several peoples with disabilities. In recent years, plant derived compounds have received much attention as potential alternatives to synthetic chemicals due

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service