Skip to Content
Merck
All Photos(1)

Key Documents

171603

Sigma-Aldrich

DL-3-Phenylserine hydrate

98%

Synonym(s):

beta-hydroxy-phenylalanine hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€112.00

€112.00


Estimated to ship on22 April 2025


Request a Bulk Order

Select a Size

Change View
25 G
€112.00

About This Item

Linear Formula:
C6H5CH(OH)CH(NH2)CO2H·xH2O
CAS Number:
Molecular Weight:
181.19 (anhydrous basis)
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

€112.00


Estimated to ship on22 April 2025


Request a Bulk Order

Quality Level

Assay

98%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

mp

186 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

O.NC(C(O)c1ccccc1)C(O)=O

InChI

1S/C9H11NO3.H2O/c10-7(9(12)13)8(11)6-4-2-1-3-5-6;/h1-5,7-8,11H,10H2,(H,12,13);1H2

InChI key

DUGXBGWTWDCGNP-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service