Skip to Content
Merck
All Photos(1)

Key Documents

171573

Sigma-Aldrich

3-Phenylpropionitrile

99%

Synonym(s):

Hydrocinnamonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€149.00

€149.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€149.00

About This Item

Linear Formula:
C6H5CH2CH2CN
CAS Number:
Molecular Weight:
131.17
Beilstein:
636348
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€149.00


Check Cart for Availability

Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.521 (lit.)

bp

113 °C/9 mmHg (lit.)

mp

−2-−1 °C (lit.)

density

1.001 g/mL at 25 °C (lit.)

functional group

nitrile
phenyl

SMILES string

N#CCCc1ccccc1

InChI

1S/C9H9N/c10-8-4-7-9-5-2-1-3-6-9/h1-3,5-6H,4,7H2

InChI key

ACRWYXSKEHUQDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The effect of 3-Phenylpropionitrile on the growth and survival of woodlouse Porcellio scaber has been studied[1].

Application

3-Phenylpropionitrile was used to study the free enzyme activity of nitrilase AtNIT1[2].

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Marijana Popović et al.
Biomolecules, 10(2) (2020-02-27)
Horseradish degradation products, mainly isothiocyanates (ITC) and nitriles, along with their precursors glucosinolates, were characterized by GC-MS and UHPLC-MS/MS, respectively. Volatiles from horseradish leaves and roots were isolated using microwave assisted-distillation (MAD), microwave hydrodiffusion and gravity (MHG) and hydrodistillation (HD).
Selective hydrolysis of the nitrile group of cis-dihydrodiols from aromatic nitriles.
Journal of Molecular Catalysis. B, Enzymatic, 38(2), 78-83 (2006)
Yu-Chun Chiu et al.
Foods (Basel, Switzerland), 9(6) (2020-06-12)
Exogenous methyl jasmonate (MeJA) treatment was known to increase the levels of neoglucobrassicin and their bioactive hydrolysis products in broccoli (Brassica oleracea var. italica), but the fate of MeJA-induced glucosinolates (GSLs) after various cooking methods was unknown. This study measured
Yu-Chun Chiu et al.
Food chemistry, 299, 125099-125099 (2019-07-13)
Applying methyl jasmonate can mimic the defense response to insect damage in broccoli and enhances the production of glucosinolates, especially inducible indolyl GS-neoglucobrassicin. Previous studies have suggested that glucosinolates and their hydrolysis products are anti-carcinogenic. Therefore, MeJA treatment may increase
A E Elaine van Ommen Kloeke et al.
Chemosphere, 89(9), 1084-1090 (2012-06-16)
Glucosinolates are compounds produced by commercial crops which can hydrolyse in a range of natural toxins that may exert detrimental effects on beneficial soil organisms. This study examined the effects of 2-phenylethyl isothiocyanate and 3-phenylpropionitrile on the survival and growth

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service