Skip to Content
Merck
All Photos(1)

Key Documents

168793

Sigma-Aldrich

Methyl 3,5-dimethoxybenzoate

99%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€171.00

€171.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€171.00

About This Item

Linear Formula:
(CH3O)2C6H3CO2CH3
CAS Number:
Molecular Weight:
196.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€171.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

solid

bp

298 °C (lit.)

mp

42-43 °C (lit.)

functional group

ester

SMILES string

COC(=O)c1cc(OC)cc(OC)c1

InChI

1S/C10H12O4/c1-12-8-4-7(10(11)14-3)5-9(6-8)13-2/h4-6H,1-3H3

InChI key

YXUIOVUOFQKWDM-UHFFFAOYSA-N

General description

Bromination of methyl 3,5-dimethoxybenzoate has been investigated[1].

Application

Methyl 3,5-dimethoxybenzoate was used as starting reagent in the synthesis of 5,7-dimethoxy-4-methylphthalide, a key intermediate in the synthesis of mycophenolic acid and its analogues[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The bromination of some derivatives of resorcinol.
Cannon JR, et al.
J. Chem. Soc. Sect. C, 3495-3504 (1971)
An Improved Synthesis of 5, 7-Dimethoxy-4-methylphthalide, A Key Intermediate in the Synthesis of Mycophenolic Acid.
Makara GM, et al.
Synthetic Communications, 26(10), 1935-1942 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service