Skip to Content
Merck
All Photos(1)

Key Documents

16772

Sigma-Aldrich

Cyanogen bromide

≥98.5% (RT)

Synonym(s):

Bromine cyanide

Sign Into View Organizational & Contract Pricing

Select a Size

200 G
€339.00

€339.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 G
€339.00

About This Item

Linear Formula:
BrCN
CAS Number:
Molecular Weight:
105.92
Beilstein:
1697296
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€339.00


Check Cart for Availability

Request a Bulk Order

vapor density

3.65 (vs air)

Quality Level

vapor pressure

100 mmHg ( 22.6 °C)

Assay

≥98.5% (RT)

form

crystals

bp

61-62 °C (lit.)

mp

50-53 °C (lit.)

functional group

bromo

storage temp.

2-8°C

SMILES string

BrC#N

InChI

1S/CBrN/c2-1-3

InChI key

ATDGTVJJHBUTRL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Cyanogen bromide is widely used in protein immobilization[1][2] and cleavage.[3]
  • Under von Braun reaction conditions, CNBr reacts with tertiary amines to yield respective organocyanamides.[4][5]
  • It can also be used to synthesize organic intermediates like cyanuric bromide, derivatives of urea, guanidine, creatine, aryl nitriles, and a variety of heterocyclic compounds.[6][7][8][9]

Other Notes

Reagent for the selective cleavage of peptide bonds (e.g. methionine)[10][11]

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

>149.0 °F - closed cup

Flash Point(C)

> 65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of hindered chiral guanidine bases starting from (S)-(N, N-dialkyl-aminomethyl) pyrrolidines and BrCN.
Kohn U, et al.
Tetrahedron Asymmetry, 17(5), 811-818 (2006)
Straightforward Conversion of Alcohols into Nitriles.
Tarrade-Matha A, et al.
Synthetic Communications, 40(11), 1646-1649 (2010)
E. Gross
Methods in Enzymology, 11, 238-238 (1967)
Immobilization of lipases on hydrophobic supports involves the open form of the enzyme.
Manoel EA, et al.
Enz. Microbiol. Technol., 71, 53-57 (2015)
The von Braun Reaction Applied to Azetidines.
Wright K, et al.
European Journal of Organic Chemistry, 2017(47), 195-7201 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service