Skip to Content
Merck
All Photos(1)

Key Documents

16571

Sigma-Aldrich

(+)-3-Bromocamphor

purum, ≥97.0% (sum of enantiomers, GC)

Synonym(s):

endo-(1R)-3-Bromo-1,7,7-trimethylbicyclo[2.2.1]heptan-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€72.70

€72.70


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
50 G
€72.70

About This Item

Empirical Formula (Hill Notation):
C10H15BrO
CAS Number:
Molecular Weight:
231.13
Beilstein:
3197237
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22

€72.70


Estimated to ship on25 May 2025


Request a Bulk Order

grade

purum

Assay

≥97.0% (sum of enantiomers, GC)

form

solid

optical activity

[α]20/D +136±3°, c = 1% in ethanol

mp

75-77 °C

functional group

bromo
ketone

SMILES string

[H][C@@]12CC[C@@](C)(C(=O)[C@H]1Br)C2(C)C

InChI

1S/C10H15BrO/c1-9(2)6-4-5-10(9,3)8(12)7(6)11/h6-7H,4-5H2,1-3H3/t6-,7+,10+/m1/s1

InChI key

NJQADTYRAYFBJN-FWWHASMVSA-N

Application

  • Insights into the mechanisms of within-species variation in sensitivity to chemicals: A case study using daphnids exposed to CMIT/MIT biocide.: This study investigates the differential sensitivity of daphnids to 2-Methyl-4-isothiazolin-3-one, highlighting the biochemical mechanisms underlying these variations. The findings provide crucial insights for environmental safety assessments (Kim et al., 2023).

Other Notes

Use as chiral building block[1][2][3]; Debromo-silylation and -phosphonylation[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P. Sampson et al.
The Journal of Organic Chemistry, 51, 4342-4342 (1986)
R.V. Stevens et al.
Tetrahedron, 41, 93-93 (1985)
J.H. Hutchinson et al.
Canadian Journal of Chemistry, 65, 1-1 (1987)
Camphor: a chiral starting material in natural product synthesis.
T Money
Natural product reports, 2(3), 253-289 (1985-06-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service