Skip to Content
Merck
All Photos(1)

Key Documents

164682

Sigma-Aldrich

Triethyloxonium hexafluorophosphate

contains diethyl ether as stabilizer

Synonym(s):

Meerwein′s salt, Triethyloxonium hexafluorophosphate(1-)

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€162.00
25 G
€592.00

€162.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€162.00
25 G
€592.00

About This Item

Linear Formula:
(C2H5)3OPF6
CAS Number:
Molecular Weight:
248.15
Beilstein:
4302183
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

€162.00


Check Cart for Availability

Request a Bulk Order

contains

diethyl ether as stabilizer

Quality Level

mp

~145 °C (dec.) (lit.)

functional group

ether

storage temp.

−20°C

SMILES string

CC[O+](CC)CC.F[P-](F)(F)(F)(F)F

InChI

1S/C6H15O.F6P/c1-4-7(5-2)6-3;1-7(2,3,4,5)6/h4-6H2,1-3H3;/q+1;-1

InChI key

JRHIFBONIQGZFC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant or reagent involved in:
  • Synthesis of ruthenium allenylidene complexes
  • Asymmetric Diels-Alder reactions
  • Chlorine scavenging
  • Synthesis of molecules for studies of wavelength tunability via cis-trans isomerization
  • Reactions with 2-mthylpyridine N-oxide

Catalyst activating agent for asymmetric imine aziridination via diazo ester activation

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service