Skip to Content
Merck
All Photos(1)

Key Documents

163732

Sigma-Aldrich

MES monohydrate

Synonym(s):

2-(N-Morpholino)ethanesulfonic acid, 4-Morpholineethanesulfonic acid monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO4S · H2O
CAS Number:
Molecular Weight:
213.25
Beilstein:
6350956
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

98.5-102.5% NaOH basis (titration)

form

powder

useful pH range

5.5 - 6.7

pKa (25 °C)

6.1

mp

>300 °C (lit.)

solubility

water: 0.25 g/mL, clear, colorless

SMILES string

O.OS(=O)(=O)CCN1CCOCC1

InChI

1S/C6H13NO4S.H2O/c8-12(9,10)6-3-7-1-4-11-5-2-7;/h1-6H2,(H,8,9,10);1H2

InChI key

MIIIXQJBDGSIKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

MES monohydrate (2-morpholinoethanesulfonic acid monohydrate) is is a zwitterionic organic compound used as an acidic buffer in specific biological processes.[1][2][3]

Application

A buffer useful in the physiological pH range.[4][5]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A buffer that mimics the SMP buffer for determining lime requirement of soil
Sikora, FJ
Soil Science Society of America Journal. Soil Science Society of America, 70(2), 474-486 (2006)
In vitro antileishmanial activity of Aloe vera leaf exudate: a potential herbal therapy in leishmaniasis
Dutta A, et al.
Glycoconjugate Journal, 24(1), 81-86 (2007)
Analytical Chemistry, 65, 247-247 (1993)
Pharmacological Research, 10, 1654-1654 (1993)
Tongtong Ma et al.
International journal of pharmaceutics, 473(1-2), 296-303 (2014-07-02)
Oral delivery of antigens is patient-friendly and efficient way of treating intestinal infections. However, the efficacy of oral vaccines is limited by degradation in the gastrointestinal (GI) tract and poor absorption by enterocytes and antigen-presenting cells (APC). Here we report

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service