Skip to Content
Merck
All Photos(2)

Key Documents

162906

Sigma-Aldrich

2-Hydroxy-5-nitrobenzyl bromide

90%

Synonym(s):

α-Bromo-4-nitro-o-cresol, 2-Bromomethyl-4-nitrophenol, Koshland I, Koshland’s Reagent I

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€59.90

€59.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€59.90

About This Item

Linear Formula:
HOC6H3(NO2)CH2Br
CAS Number:
Molecular Weight:
232.03
Beilstein:
1868798
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€59.90


Check Cart for Availability

Request a Bulk Order

Assay

90%

form

solid

mp

144-149 °C (lit.)

solubility

chloroform: soluble 25 mg/mL, clear, yellow

functional group

bromo
nitro

storage temp.

2-8°C

SMILES string

Oc1ccc(cc1CBr)[N+]([O-])=O

InChI

1S/C7H6BrNO3/c8-4-5-3-6(9(11)12)1-2-7(5)10/h1-3,10H,4H2

InChI key

KFDPCYZHENQOBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Hydroxy-5-nitrobenzyl bromide is a protein modifying reagent[1].

Application

2-Hydroxy-5-nitrobenzyl bromide was used in covalent modification of tryptophan[2] and tryptophan residues in monoclonal immunoglobulin[3]. It was also used as reagent for sulfhydryl modification[4]

Other Notes

Contains 2-hydroxy-5-nitrobenzyl alcohol

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Solaini et al.
The Biochemical journal, 327 ( Pt 2), 443-448 (1997-11-14)
Treatment of bovine heart submitochondrial particles with a low concentration of 2-hydroxy-5-nitrobenzyl bromide (HNB), a selective reagent for the Trp residue of the epsilon subunit [Baracca, Barogi, Lenaz and Solaini (1993) Int. J. Biochem. 25, 1269-1275], enhances the ATP hydrolytic
A Baracca et al.
Biochemistry international, 21(6), 1135-1142 (1990-09-01)
The incubation of bovine mitochondrial F1-ATPase with 2-hydroxy-5-nitrobenzyl bromide (HNB), a selective reagent toward tryptophan residues in proteins, produced a concentration dependent inactivation of the enzyme and the covalent binding of 0.88 mol reagent/mol F1. Although HNB is highly specific
S S Keskar et al.
The Biochemical journal, 261(1), 49-55 (1989-07-01)
Extracellular xylanase produced in submerged culture by a thermotolerant Streptomyces T7 growing at 37-50 degrees C was purified to homogeneity by chromatography on DEAE-cellulose and gel filtration on Sephadex G-50. The purified enzyme has an Mr of 20,463 and a
V A Lapuk et al.
Biokhimiia (Moscow, Russia), 50(2), 237-242 (1985-02-01)
The modification of tryptophan residues in monoclonal immunoglobulin M (IgM) by 2-hydroxy-5-nitrobenzyl bromide (RK) was studied at pH 2.0-2.85 and 7.0 and a RK to tryptophan molar ratio (K) from 1 to 40. At pH 2.85, the number of RK
A I Korneliuk et al.
Bioorganicheskaia khimiia, 11(5), 605-612 (1985-05-01)
The structural accessibility of tryptophan residues in leucyl-tRNA synthetase from cow mammary gland has been studied using chemical modifications by N-bromosuccinimide and 2-hydroxy-5-nitrobenzyl bromide. The modifications were monitored by UV absorbance and intrinsic fluorescence of the enzyme's tryptophan residues. Under

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service