Skip to Content
Merck
All Photos(2)

Key Documents

162620

Sigma-Aldrich

N-Z-L-aspartic acid

99%

Synonym(s):

Z-L-aspartic acid, Z-Asp-OH

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€71.50

€71.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€71.50

About This Item

Linear Formula:
HOOCCH2CH(NHCOOCH2C6H5)COOH
CAS Number:
Molecular Weight:
267.23
Beilstein:
2336722
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

€71.50


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

solid

optical activity

[α]23/D +8.6°, c = 7 in acetic acid

reaction suitability

reaction type: solution phase peptide synthesis

mp

117-119 °C (lit.)

application(s)

peptide synthesis

SMILES string

OC(=O)C[C@H](NC(=O)OCc1ccccc1)C(O)=O

InChI

1S/C12H13NO6/c14-10(15)6-9(11(16)17)13-12(18)19-7-8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,13,18)(H,14,15)(H,16,17)/t9-/m0/s1

InChI key

XYXYXSKSTZAEJW-VIFPVBQESA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xilong Wu et al.
Advanced healthcare materials, 7(11), e1800121-e1800121 (2018-03-28)
Autophagy is a homeostatic lysosome-dependent metabolic process to eliminate damaged or dysfunctional cellular organelles, which is closely associated with tumor progression. Indocyanine green (ICG) can convert NIR light energy to localized heat for cancer cell and tissue ablation. However, the
Griet Van Zeebroeck et al.
Nature chemical biology, 5(1), 45-52 (2008-12-09)
Transporter-related nutrient sensors, called transceptors, mediate nutrient activation of signaling pathways through the plasma membrane. The mechanism of action of transporting and nontransporting transceptors is unknown. We have screened 319 amino acid analogs to identify compounds that act on Gap1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service