Skip to Content
Merck
All Photos(1)

Key Documents

162299

Sigma-Aldrich

1-Chloro-3-fluorobenzene

99%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€87.20

€87.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€87.20

About This Item

Linear Formula:
ClC6H4F
CAS Number:
Molecular Weight:
130.55
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€87.20


Check Cart for Availability

Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.494 (lit.)

bp

126-128 °C (lit.)

density

1.219 g/mL at 25 °C (lit.)

functional group

chloro
fluoro

SMILES string

Fc1cccc(Cl)c1

InChI

1S/C6H4ClF/c7-5-2-1-3-6(8)4-5/h1-4H

InChI key

VZHJIJZEOCBKRA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Reaction of 1-chloro-3-fluorobenzene radical cation with NH3 has been investigated by FT-ICR spectrometry[1]. It reacts with n-butyllithium to yield bicycloadducts via trapping of intermediate benzynes[2].

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactions of dihalobenzene radical cations with ammonia in the gas phase. Reactivity pattern for nucleophilic aromatic substitution.
Tholmann D and Gruetzmacher HF.
Journal of the American Chemical Society, 113(9), 3281-3287 (1991)
Antonio Ramírez et al.
Journal of the American Chemical Society, 126(45), 14700-14701 (2004-11-13)
The key elimination step for the formation of 3-chloro- and 3-fluorobenzyne from 2-chloro-6-fluorophenyllithium displays a pronounced solvent-dependent regioselectivity. 6Li and 13C NMR spectroscopic studies on 2-chloro-6-fluorophenyllithium reveal a single monomeric aryllithium, suggested by DFT computational studies to be a trisolvate.
Oriol Planas et al.
Science (New York, N.Y.), 367(6475), 313-317 (2020-01-18)
Bismuth catalysis has traditionally relied on the Lewis acidic properties of the element in a fixed oxidation state. In this paper, we report a series of bismuth complexes that can undergo oxidative addition, reductive elimination, and transmetallation in a manner

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service