Skip to Content
Merck
All Photos(1)

Key Documents

162108

Sigma-Aldrich

4-Aminoindan

97%

Synonym(s):

4-Indanamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€57.60

€57.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€57.60

About This Item

Empirical Formula (Hill Notation):
C9H11N
CAS Number:
Molecular Weight:
133.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€57.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.592 (lit.)

bp

90-92 °C (lit.)

density

1.102 g/mL at 25 °C (lit.)

SMILES string

Nc1cccc2CCCc12

InChI

1S/C9H11N/c10-9-6-2-4-7-3-1-5-8(7)9/h2,4,6H,1,3,5,10H2

InChI key

RXTJLDXSGNEJIT-UHFFFAOYSA-N

General description

4-Aminoindan undergoes nucleophlic addition reaction with 2-chloroacetylisocyanate to yield 1-(2-chloroacetyl)-3-(2,3-dihydro-1H-inden-4-yl)-urea[1].

Application

4-Aminoindan was used in the synthesis of 4,5-dihydro-N-(2,3-dihydro-1H-inden-4-yl)oxazol-2-amine[1] and 1-(2-chloroethyl)-3-(2,3-dihydro-1H-inden-4-yl)urea[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica S Fortin et al.
Bioorganic & medicinal chemistry, 15(13), 4456-4469 (2007-05-15)
A number of N-phenyl-N'-(2-chloroethyl)ureas (CEUs) have been shown to be potent antimitotics through their covalent binding to the colchicine-binding site on intracellular beta-tubulin. The present communication aimed to evaluate the role of the electrophilic 2-chloroethyl amino moiety of CEU on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service