Skip to Content
Merck
All Photos(1)

Key Documents

161438

Sigma-Aldrich

2-Chloroanisole

98%

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€72.90

€72.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€72.90

About This Item

Linear Formula:
ClC6H4OCH3
CAS Number:
Molecular Weight:
142.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€72.90


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.545 (lit.)

bp

195-196 °C (lit.)

density

1.123 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

COc1ccccc1Cl

InChI

1S/C7H7ClO/c1-9-7-5-3-2-4-6(7)8/h2-5H,1H3

InChI key

QGRPVMLBTFGQDQ-UHFFFAOYSA-N

Related Categories

General description

2-Chloroanisole undergoes acetylation with acetic anhydride over large pore zeolites to give 4-acetyl-2-chloroanisole[1].

Application

2-Chloroanisole was used in headspace solid-phase microextraction method for the determination of haloanisoles in wine and spirit samples[2].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Keith Smith et al.
Organic & biomolecular chemistry, 1(9), 1560-1564 (2003-08-21)
The acetylation of aryl ethers using acetic anhydride in the presence of zeolites under modest conditions in a solvent-free system gave the corresponding para-acetylated products in high yields. The zeolite can be recovered, regenerated and reused to give almost the
Natalia Campillo et al.
Journal of chromatography. A, 1210(2), 222-228 (2008-10-18)
A headspace solid-phase microextraction (HS-SPME) method for the determination of 12 haloanisoles in wine and spirit samples using gas chromatography with atomic emission detection (GC-AED) was developed. The different factors affecting the efficiency of the extraction were carefully optimized. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service