Skip to Content
Merck
All Photos(1)

Key Documents

16125

Sigma-Aldrich

Ethyl cis-3-bromoacrylate

≥99.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7BrO2
CAS Number:
Molecular Weight:
179.01
Beilstein:
1902817
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (GC)

refractive index

n20/D 1.481

density

1.48 g/mL at 20 °C

functional group

bromo
ester

storage temp.

2-8°C

SMILES string

CCOC(=O)\C=C/Br

InChI

1S/C5H7BrO2/c1-2-8-5(7)3-4-6/h3-4H,2H2,1H3/b4-3-

InChI key

UJTJVQIYRQALIK-ARJAWSKDSA-N

Application

Ethyl cis-3-bromoacrylate was used in the preparation of ethyl 2-amino-9-(2-deoxy-β-D-erythropentofuranosyl)-6,9-dihydro-6-oxo-1H-purine-1-acrylate.[1] It was used in the synthesis of 3-(2-deoxy-β-D-erythro-pentofuranosyl)-3,4-dihydropyrimido[1,2-a]purine-6,10-dione.[2] It was employed as building block for the stereoselective preparation of cis-2-enoates.[3][4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A.B. Smith et al.
Tetrahedron Letters, 26, 4419-4419 (1985)
X. Lu et al.
Tetrahedron Letters, 33, 2535-2535 (1992)
Jozsef Szekely et al.
Nucleosides, nucleotides & nucleic acids, 27(2), 103-109 (2008-01-22)
Short, "one-pot" syntheses of malondialdehyde adducts of deoxyguanosine, deoxyadenosine, and deoxycytidine are described. These syntheses proceed in improved yield and easier purification than previous syntheses and are well suited for the preparation of isotopically labeled nucleoside adducts for biomarker and
Michael B Otteneder et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(17), 6665-6669 (2006-04-15)
3-(2-Deoxy-beta-D-erythro-pentofuranosyl)pyrimido[1,2-alpha]purin-10(3H)-one (M1dG) is a DNA adduct arising from the reaction of 2-deoxyguanosine with the lipid peroxidation product, malondialdehyde, or the DNA peroxidation product, base propenal. M1dG is mutagenic in bacteria and mammalian cells and is present in the genomic DNA
Alessio Grimaldi et al.
Communications biology, 3(1), 85-85 (2020-02-27)
Here, we developed an unbiased, functional target-discovery platform to identify immunogenic proteins from primary non-small cell lung cancer (NSCLC) cells that had been induced to apoptosis by cisplatin (CDDP) treatment in vitro, as compared with their live counterparts. Among the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service