Skip to Content
Merck
All Photos(1)

Key Documents

159913

Sigma-Aldrich

Heptanophenone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CO(CH2)5CH3
CAS Number:
Molecular Weight:
190.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.5077 (lit.)

bp

155 °C/15 mmHg (lit.)

mp

17 °C (lit.)

density

0.946 g/mL at 25 °C (lit.)

SMILES string

CCCCCCC(=O)c1ccccc1

InChI

1S/C13H18O/c1-2-3-4-8-11-13(14)12-9-6-5-7-10-12/h5-7,9-10H,2-4,8,11H2,1H3

InChI key

UXMQORVHJMUQFD-UHFFFAOYSA-N

General description

Heptanophenone is an alkyl phenyl ketone and its inhibitory effect on carbonyl reductase activity in pig heart has been examined.

Application

Heptanophenone was employed as test compound to compare the kinetic performance of monolithic and fused-core capillary C(18) columns in isocratic-elution liquid chromatography.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sam Wouters et al.
Journal of chromatography. A, 1523, 224-233 (2017-06-18)
The present paper discusses practical aspects of prototyping of microfluidic chips using cyclic olefin copolymer as substrate and the application in high-performance liquid chromatography. The developed chips feature a 60mm long straight separation channel with circular cross section (500μm i.d.)
Tünde A Diószegi et al.
Journal of chromatography. A, 1261, 107-112 (2012-09-12)
The kinetic performance of monolithic and fused-core capillary C(18) columns was compared in isocratic-elution liquid chromatography. Heptanophenone was chosen as a test compound. The (u(0), H) couple obtained in a column with permeability K(v0) was transferred into a plate number
Shunta Futagami et al.
The Analyst, 144(5), 1809-1817 (2019-01-24)
The performance of a porous-layered radially elongated pillar (PLREP) array column in a commercial nano-LC system was examined by performing separation of alkylphenones and peptides. The mesoporous silica layer was prepared by sol-gel processing of a mixture of tetramethoxysilane and
Gert Desmet et al.
Journal of chromatography. A, 1626, 461279-461279 (2020-08-17)
We report on a comparative study of the basic separation kinetics of commercial packed bed columns and a micro-pillar array column (μPAC) working in the 1-10μL/min flow rate range, i.e., operating in the area of capillary flow LC. This is
Erin P Shields et al.
Journal of chromatography. A, 1598, 132-140 (2019-04-15)
Stationary phases that can withstand extremes of pH and temperature are needed to allow a single column to accommodate a wider set of solutes and separation criteria. We used a simple multi-step process using the thiol-yne reaction following the modification

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service