Skip to Content
Merck
All Photos(2)

Documents

159344

Sigma-Aldrich

Ethyl 4-isocyanatobenzoate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OCNC6H4CO2C2H5
CAS Number:
Molecular Weight:
191.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

bp

118-119 °C/0.8 mmHg (lit.)

mp

27-29 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)c1ccc(cc1)N=C=O

InChI

1S/C10H9NO3/c1-2-14-10(13)8-3-5-9(6-4-8)11-7-12/h3-6H,2H2,1H3

InChI key

CFEPCPHKICBCJV-UHFFFAOYSA-N

Application

Ethyl 4-isocyanatobenzoate was used in the preparation of cellulose carbamate and ester derivatives. It was also used in the preparation of ethyl 4-(3-(4-oxo-6-tridecyl-1,4-dihydropyrimidin-2-yl)ureido)benzoate and ethyl 4-(2-oxocyclohexanecarboxamido)benzoate.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cellulose derivatives synthesized via isocyanate and activated ester pathways in homogeneous solutions of lithium chloride/N, N-dimethylacetamide.
Williamson SL and McCormick CL.
J. Macromol. Sci., Pure Appl. Chem., 35(12), 1915-1927 (1998)
Chun-Ho Park et al.
Bioorganic & medicinal chemistry letters, 20(7), 2250-2253 (2010-03-02)
Highly potent poly(ADP-ribose)polymerase-1 (PARP-1) inhibitors, including 9-hydroxy-1,2-dihydro-4H-thiopyrano[3,4-c]quinolin-5(6H)-one derivatives with a non-aromatic A-ring, were synthesized. Among the derivatives, 12a showed low nanomolar enzyme and cellular activity (IC(50) = 42 nM, ED(50) = 220 nM) with good water solubility. Further, 12a exhibited
Electronic substituent effects on hydrogen-bonding motifs modulate supramolecular polymerisation.
Pellizzaro ML, et al.
Royal Society of Chemistry Advances, 3(9), 3103-3108 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service