Skip to Content
Merck
All Photos(1)

Key Documents

159182

Sigma-Aldrich

Ethyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO2S
CAS Number:
Molecular Weight:
225.31
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

114-116 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)c1c(N)sc2CCCCc12

InChI

1S/C11H15NO2S/c1-2-14-11(13)9-7-5-3-4-6-8(7)15-10(9)12/h2-6,12H2,1H3

InChI key

CDYVTVLXEWMCHU-UHFFFAOYSA-N

Gene Information

human ... ADORA1(134)

Application

Ethyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate was used in the preparation of:
  • thienopyrimidine derivatives[1]
  • azo dyes[2]
  • N-(3-ethoxycarbonyl-4,5,6,7-tetrahydrobenzo[b]thien-2-yl)-2-aryl hydrazono-3-oxobutanamide derivatives[3]
  • tetrahydrobenzo[b]thiophene dyes[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M M el-Kerdawy et al.
Bollettino chimico farmaceutico, 135(5), 301-305 (1996-05-01)
Interaction of ethyl 2-amino-4,5,6, 7-tetrahydrobenzo[b]thiophene-3-carboxylate 1 with 2-thiophene carbonyl chloride 2 afforded ethyl 2-(2-thenoylamino)-4, 5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate 3 which upon cyclization yielded 2-(2-thienyl)-5,6,7, 8-tetrahydrobenzo[b]-thieno[2,3-d]-4H-3, 1-oxazin-4-one 4 and 2(2-thienyl)-3-amino-5, 6,7,8-tetrahydrobenzo-[b]thieno[2,3-d]-3,4-dihydropyrimidin-4-one 6. Some of the prepared compounds were screened for their antiinflammatory activity, compound
Synthesis, azo-hydrazone tautomerism and antitumor screening of N-(3-ethoxycarbonyl-4, 5, 6, 7-tetrahydro-benzo [b] thien-2-yl)-2-arylhydrazono-3-oxobutanamide derivatives.
Farghaly TA and Abdallah ZA.
ARKIVOC (Gainesville, FL, United States), 17, 295-305 (2008)
Synthesis of azobenzo [b] thiophene derivatives and their dyeing performance on polyester fibre
Maradiya HR.
Turkish Journal of Chemistry, 25(4), 441-450 (2001)
Synthesis of azo benzo [b] thiophene derivatives and their application as disperse dyes.
Sabnis RW and Rangnekar DW.
Dyes and Pigments, 10(4), 295-302 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service