Skip to Content
Merck
All Photos(1)

Key Documents

156825

Sigma-Aldrich

Boron trifluoride propanol complex

in excess propanol, BF3 14 wt. %

Synonym(s):

(T-4)-Trifluoro(1-propanol)boron, Boron trifluoride-propanol complex (1:1)

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€129.00

€129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€129.00

About This Item

Linear Formula:
BF3 · CH3CH2CH2OH
CAS Number:
Molecular Weight:
127.90
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

€129.00


Check Cart for Availability

Request a Bulk Order

form

liquid

composition

BF3, 14 wt. %

reaction suitability

core: boron
reagent type: Lewis acid
reagent type: catalyst

concentration

in excess propanol

refractive index

n20/D 1.379 (lit.)

density

0.905 g/mL at 25 °C (lit.)

SMILES string

CCCO.FB(F)F

InChI

1S/C3H8O.BF3/c1-2-3-4;2-1(3)4/h4H,2-3H2,1H3;

InChI key

RQPQOULISUUGNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service