Skip to Content
Merck
All Photos(2)

Key Documents

155403

Sigma-Aldrich

4-Nitrophenyl phosphorodichloridate

98%

Synonym(s):

4-Nitrophenyl dichlorophosphate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€90.40
25 G
€263.00

€90.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€90.40
25 G
€263.00

About This Item

Linear Formula:
O2NC6H4OP(O)Cl2
CAS Number:
Molecular Weight:
255.98
Beilstein:
1468239
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€90.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

bp

182-183 °C/13 mmHg (lit.)

mp

44-45 °C (lit.)

functional group

nitro

SMILES string

[O-][N+](=O)c1ccc(OP(Cl)(Cl)=O)cc1

InChI

1S/C6H4Cl2NO4P/c7-14(8,12)13-6-3-1-5(2-4-6)9(10)11/h1-4H

InChI key

NCPBESHYZRJICR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Nitrophenyl phosphorodichloridate was used to phosphorylate the derivatized resin[1].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

An enzyme-scissile linker for solid-phase peptide synthesis.
Elmore DT, et al. et al.
Journal of the Chemical Society. Chemical Communications, 14, 1033-1034 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service