Skip to Content
Merck
All Photos(1)

Key Documents

15533

Sigma-Aldrich

Boc-Abu-OH

≥97.0% (HPLC)

Synonym(s):

(S)-2-(Boc-amino)butyric acid, Boc-L-2-aminobutyric acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€220.00

€220.00


Estimated to ship on18 April 2025


Request a Bulk Order

Select a Size

Change View
10 G
€220.00

About This Item

Linear Formula:
C2H5CH[NHCO2C(CH3)3]CO2H
CAS Number:
Molecular Weight:
203.24
Beilstein:
6801706
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

€220.00


Estimated to ship on18 April 2025


Request a Bulk Order

Quality Level

Assay

≥97.0% (HPLC)

optical activity

[α]20/D −18.5±2°, c = 1% in methanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

application(s)

peptide synthesis

SMILES string

CC[C@H](NC(=O)OC(C)(C)C)C(O)=O

InChI

1S/C9H17NO4/c1-5-6(7(11)12)10-8(13)14-9(2,3)4/h6H,5H2,1-4H3,(H,10,13)(H,11,12)/t6-/m0/s1

InChI key

PNFVIPIQXAIUAY-LURJTMIESA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fahd Boutouja et al.
Biomolecules, 10(11) (2020-11-19)
The changing accessibility of nutrient resources induces the reprogramming of cellular metabolism in order to adapt the cell to the altered growth conditions. The nutrient-depending signaling depends on the kinases mTOR (mechanistic target of rapamycin), which is mainly activated by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service