Skip to Content
Merck
All Photos(5)

Key Documents

155322

Sigma-Aldrich

trans-Styrylacetic acid

greener alternative

96%

Synonym(s):

4-Phenyl-3-butenoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€48.40
10 G
€197.00

€48.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€48.40
10 G
€197.00

About This Item

Linear Formula:
C6H5CH=CHCH2CO2H
CAS Number:
Molecular Weight:
162.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€48.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

greener alternative product score

old score: 22
new score: 3
Find out more about DOZN™ Scoring

greener alternative product characteristics

Atom Economy
Design for Energy Efficiency
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

84-86 °C (lit.)

functional group

carboxylic acid
phenyl

greener alternative category

SMILES string

OC(=O)C\C=C\c1ccccc1

InChI

1S/C10H10O2/c11-10(12)8-4-7-9-5-2-1-3-6-9/h1-7H,8H2,(H,11,12)/b7-4+

InChI key

PSCXFXNEYIHJST-QPJJXVBHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The polarized IR spectra of the hydrogen bond system in trans-styrylacetic acid has been studied[1].
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Atom Economy”, “Design for Energy Efficiency” and “Use of Renewable Feedstock”. Click here to view its DOZN scorecard.

Application

trans-Styrylacetic acid (4-Phenyl-3-butenoic acid) was used as mechanism-based inhibitor of Peptidylglycine α-hydroxylating monooxygenase[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Scognamiglio et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50 Suppl 2, S120-S123 (2011-10-29)
A toxicologic and dermatologic review of 4-phenyl-3-buten-2-ol when used as a fragrance ingredient is presented. 4-Phenyl-3-buten-2-ol is a member of the fragrance structural group Aryl Alkyl Alcohols and is a secondary alcohol. The AAAs are a structurally diverse class of
Henryk T Flakus et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 65(2), 481-489 (2006-03-11)
We have investigated the polarized IR spectra of the hydrogen bond system in crystals of trans-styrylacetic acid C(6)H(5)CHCHCH(2)COOH, and also in crystals of the following three deuterium isotopomers of the compound: C(6)H(5)CHCHCH(2)COOD, C(6)H(5)CHCHCD(2)COOH and C(6)H(5)CHCHCD(2)COOD. The spectra were measured at
David J Merkler et al.
Biochemistry, 43(39), 12667-12674 (2004-09-29)
Oleamide is an endogenous sleep-inducing lipid that has been isolated from the cerebrospinal fluid of sleep-deprived mammals. Oleamide is the best-understood member of the primary fatty acid amide family. One key unanswered question regarding oleamide and all other primary acid
W J Driscoll et al.
Biochemistry, 39(27), 8007-8016 (2000-07-13)
The bifunctional enzyme peptidylglycine-alpha-amidating monooxygenase mediates the conversion of C-terminal glycine-extended peptides to their active alpha-amidated products. Peptidylglycine-alpha-hydroxylating monooxygenase (PHM, EC 1.14.17. 3) catalyzes the first reaction in this two-step process. The olefinic compound 4-phenyl-3-butenoic acid (PBA) is the most
Yuan-Ning Cao et al.
Regulatory peptides, 113(1-3), 109-114 (2003-04-11)
Human adrenomedullin (hAM) is an endogenous peptide that has potent vasodilator activity. Mature AM is biosynthesized from its intermediate form, glycine-extended AM (AM-gly), by carboxy-terminal amidation. AM-gly is generally considered to be biologically inactive but is a major molecular form

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service