Skip to Content
Merck
All Photos(2)

Key Documents

155306

Sigma-Aldrich

N1-(6-Indazolyl)sulfanilamide

98%

Synonym(s):

6-Sulfanilamidoindazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12N4O2S
CAS Number:
Molecular Weight:
288.32
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

193-195 °C (lit.)

SMILES string

Nc1ccc(cc1)S(=O)(=O)Nc2ccc3cn[nH]c3c2

InChI

1S/C13H12N4O2S/c14-10-2-5-12(6-3-10)20(18,19)17-11-4-1-9-8-15-16-13(9)7-11/h1-8,17H,14H2,(H,15,16)

InChI key

RLNLIVBLEZDLMZ-UHFFFAOYSA-N

General description

N1-(6-Indazolyl)sulfanilamide ( 6-Sulfanilamidoindazole) causes inflammation in the hind paws and ankles of older rat and sensitizes them to the lethal action of endotoxin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Hirschelmann et al.
Agents and actions, 10(5), 431-434 (1980-11-01)
During 6-sulphanilamidoindazole (6-SAI) arthritis a significant increase of serum levels of the protease inhibitor alpha 2 macroglobulin (= alpha 2 acute phase globulin) and of Darcy's glycoprotein was found, whereas the content of female specific protein and of sulfhydryl groups
D D Blatter
Skeletal radiology, 16(3), 183-189 (1987-01-01)
Phosphorus-31 magnetic resonance spectroscopy (MRS) of the hind paws of rats was performed at 1.5 Tesla before and during the course of an experimentally-induced inflammatory arthritis. Arthritis was induced by daily subcutaneous administration of 6-sulfanilamidoindazole, an antibacterial sulfa known to
R Hirschelmann et al.
International journal of tissue reactions, 8(6), 469-473 (1986-01-01)
Arthritis induced by 6-sulphanilamidoindazole in older rats is not inhibited by a one-week treatment with superoxide dismutase, peroxidase or desferrioxamine. It is concluded that oxygen radicals hardly play a special part in the pathogenesis of 6-SAI arthritis. C-reactive protein and
D Müller et al.
Agents and actions, 11(6-7), 736-740 (1981-12-01)
Hexobarbital sleeping time was prolonged and ethylmorphine N-demethylation was inhibited after a single dosage or seven administration of 6-SAI to old rats. These effects were independent of the development of arthritis. Changes in cytochrome P-450 concentration after 6-SAI treatment were
Y Ohmachi et al.
Toxicologic pathology, 26(2), 262-270 (1998-04-21)
6-Sulfanilamidoindazole (6SAI) is a sulfonamide that induces acute, self-limiting arthritis in rats, and 6SAI-induced arthritis is thought to be a model for testing anti-inflammatory agents. In this study, in order to clarify the location of arthritis and relationships between arthritis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service