Skip to Content
Merck
All Photos(1)

Key Documents

155241

Sigma-Aldrich

1-Methyl-2-piperidinemethanol

98%

Synonym(s):

2-Hydroxymethyl-1-methylpiperidine, NSC 45464

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NO
CAS Number:
Molecular Weight:
129.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.4823 (lit.)

bp

79-80 °C/7 mmHg (lit.)

density

0.984 g/mL at 25 °C (lit.)

SMILES string

CN1CCCCC1CO

InChI

1S/C7H15NO/c1-8-5-3-2-4-7(8)6-9/h7,9H,2-6H2,1H3

InChI key

HXXJMMLIEYAFOZ-UHFFFAOYSA-N

General description

The standard molar energy of combustion of 1-Methyl-2-piperidinemethanol has been studied[1].

Application

Reactant for synthesis of:
Selective MCH1R antagonists
Human GnRH receptor antagonists
Dual vascular endothelial growth factor receptor-2 and fibroblast growth factor receptor 1 inhibitors
Radioiodinatable brain imaging agents for CB1 cannabinoid receptor
GABAA receptor agonists at the α3 subunit
Carbamates and N-alkylimidazoles from reaction of carbonyldiimidazoles with alcohols

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Standard molar enthalpies of formation of 1-methyl-2-piperidinemethanol, 1-piperidineethanol, and 2-piperidineethanol.
Ribeiro da Silva, et al.
The Journal of Chemical Thermodynamics, 38(11), 1461-1466 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service