Skip to Content
Merck
All Photos(1)

Key Documents

15523

Sigma-Aldrich

N-Boc-4-isothiocyanatobutylamine

≥97.0%

Synonym(s):

tert-Butyl N-(4-isothiocyanatobutyl)carbamate, Boc-4-aminobutyl isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18N2O2S
CAS Number:
Molecular Weight:
230.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97.0%

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.504

density

1.067 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)NCCCCN=C=S

InChI

1S/C10H18N2O2S/c1-10(2,3)14-9(13)12-7-5-4-6-11-8-15/h4-7H2,1-3H3,(H,12,13)

InChI key

KWATYFKNNFFFSH-UHFFFAOYSA-N

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ioannis Tsogas et al.
Chembiochem : a European journal of chemical biology, 8(15), 1865-1876 (2007-09-15)
The ability of guanidinylated poly(propylene imine) dendrimers to translocate across lipid bilayers was assessed by employing either a model phosphate-bearing liposomal membrane system or A549 human lung carcinoma cells. Two dendrimer generations, differing in the number of surface guanidinium groups
Jessica I Selig et al.
Biochimica et biophysica acta. Molecular basis of disease, 1865(9), 2526-2537 (2019-06-04)
Type 2 diabetes is a known risk factor for cardiovascular diseases and is associated with an increased risk to develop aortic heart valve degeneration. Nevertheless, molecular mechanisms leading to the pathogenesis of valve degeneration in the context of diabetes are
Alexander August et al.
Journal of neurochemistry, 151(5), 626-641 (2019-05-08)
The amyloid precursor protein (APP) and its homologs amyloid precursor-like protein 1 (APLP1) and APLP2 have central physiological functions in transcellular adhesion that depend on copper and zinc mediated trans-directed dimerization of the extracellular domains E1 and E2. Copper binds to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service