Skip to Content
Merck
All Photos(1)

Key Documents

155152

Sigma-Aldrich

Bis(4-chlorophenoxy)acetic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ClC6H4O)2CHCO2H
CAS Number:
Molecular Weight:
313.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

mp

138-140 °C (lit.)

SMILES string

OC(=O)C(Oc1ccc(Cl)cc1)Oc2ccc(Cl)cc2

InChI

1S/C14H10Cl2O4/c15-9-1-5-11(6-2-9)19-14(13(17)18)20-12-7-3-10(16)4-8-12/h1-8,14H,(H,17,18)

InChI key

ZKNSZZXBPSICFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A N Kondrat'ev et al.
Vestnik khirurgii imeni I. I. Grekova, 166(6), 56-60 (2008-04-17)
The investigation has shown that solutions of hydroxyethyl starch included in complex infusion therapy facilitate stabilization of hemodynamics in neurosurgical patients during surgery. Refortan exerts a more pronounced effect on the indices of hemodynamics as compared with voluven, but its
Michael Winterhalter et al.
European journal of medical research, 15(9), 383-389 (2010-10-19)
the constantly growing amount of different kinds of colloid fluids necessitates comparative investigations with regards to the safety and effectivity in clinical use of these preparations. Hence we compared three colloid fluids in an observational study. The objective was the
V V Moskalev et al.
Anesteziologiia i reanimatologiia, (1)(1), 32-35 (2005-04-21)
The paper comparatively evaluates the efficiency of the use of physiological sodium chloride solution, freshly-frozen plasma, 6% and 10% solutions of hydroxyethyl starch to compensate for circulating blood volume deficit in 80 neonatal infants aged 1-3 days who had hypovolemia
A N Kondrat'ev et al.
Anesteziologiia i reanimatologiia, (2)(2), 27-30 (2008-06-11)
The paper provides the results of a complex experimental and clinical study of the effects of infusion solutions of hydroxyethyl starch (HES 200/0.5--Refortan and HES 130/0.4--Voluven) on hemostatic and systemic hemodynamic parameters in patients operated on for neurosurgical pathology of
V O Kryliuk
Klinichna khirurhiia, (2)(2), 67-70 (2013-05-28)
The peculiarities of course of endogenous intoxication (EI) in severe combined abdominal trauma were studied up in experiment. There was established, that in the laboratory animals, to whom the blood loss compensation was done, using colloid-hyperosmolar solution, the El severity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service