Skip to Content
Merck
All Photos(1)

Key Documents

154113

Sigma-Aldrich

4-Methoxy-α-methylbenzyl alcohol

99%

Synonym(s):

(4-Methoxyphenyl)methylcarbinol, (±)-1-(4-Methoxyphenyl)-1-ethanol, (±)-1-(4-Methoxyphenyl)ethanol, (±)-1-(p-Methoxyphenyl)ethanol, 1-(4-Methoxyphenyl)ethan-1-ol

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€281.00

€281.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
€281.00

About This Item

Linear Formula:
CH3OC6H4CH(CH3)OH
CAS Number:
Molecular Weight:
152.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€281.00


Check Cart for Availability

Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.533 (lit.)

bp

95 °C/1 mmHg (lit.)

density

1.079 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

COc1ccc(cc1)C(C)O

InChI

1S/C9H12O2/c1-7(10)8-3-5-9(11-2)6-4-8/h3-7,10H,1-2H3

InChI key

IUUULXXWNYKJSL-UHFFFAOYSA-N

Application

4-Methoxy-α-methylbenzyl alcohol was used to study the steady-state and nanosecond, laser-flash photolysis[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tiziana Del Giacco et al.
Physical chemistry chemical physics : PCCP, 10(1), 200-210 (2007-12-14)
Steady-state and nanosecond laser flash photolysis measurements of 4-methoxybenzyl alcohol (1a), 4-methoxy-alpha-methylbenzyl alcohol (1b), 4,4'-dimethoxydiphenylmethanol (1c) and 4-methoxy-alpha,alpha'-dimethylbenzyl alcohol (1d) were carried out in air-equilibrated CH(2)Cl(2) and CH(3)CN solutions, in the presence of 9,10-dicyanoanthracene (DCA) and N-methylquinolinium tetrafluoroborate (NMQ(+)BF(4)(-)) as
Fanyu Zhang et al.
Nature communications, 11(1), 1431-1431 (2020-03-20)
The production of 2D metal-organic frameworks (MOFs) with highly exposed active surfaces is of great importance for catalysis. Here we demonstrate the formation of MOF nanosheets by utilizing CO2 as a capping agent to control the oriented growth of MOF. This
Xiao-Hong Chen et al.
PloS one, 9(4), e94543-e94543 (2014-04-18)
A novel carbonyl reductase (AcCR) catalyzing the asymmetric reduction of ketones to enantiopure alcohols with anti-Prelog stereoselectivity was found in Acetobacter sp. CCTCC M209061 and enriched 27.5-fold with an overall yield of 0.4% by purification. The enzyme showed a homotetrameric

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service