Skip to Content
Merck
All Photos(3)

Key Documents

153842

Sigma-Aldrich

4-tert-Amylphenol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C(CH3)2C6H4OH
CAS Number:
Molecular Weight:
164.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

bp

255 °C (lit.)

mp

88-89 °C (lit.)

solubility

H2O: insoluble
alcohols: soluble
benzene: soluble
chloroform: soluble
diethyl ether: soluble

SMILES string

CCC(C)(C)c1ccc(O)cc1

InChI

1S/C11H16O/c1-4-11(2,3)9-5-7-10(12)8-6-9/h5-8,12H,4H2,1-3H3

InChI key

NRZWYNLTFLDQQX-UHFFFAOYSA-N

Gene Information

rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

General description

A high performance liquid chromatography (HPLC) approach was studied for the determination of 4-tert-amylphenol[1].

Application

4-tert-Amylphenol was used as an esterogen receptor (ER) ligand[2].

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service