Skip to Content
Merck
All Photos(1)

Documents

151327

Sigma-Aldrich

2,5-Dimethyl-3-pyrroline, mixture of cis and trans

90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11N
CAS Number:
Molecular Weight:
97.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

refractive index

n20/D 1.4404 (lit.)

bp

102-105 °C (lit.)

density

0.824 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1NC(C)C=C1

InChI

1S/C6H11N/c1-5-3-4-6(2)7-5/h3-7H,1-2H3

InChI key

TXQDHQBSNAJSHQ-UHFFFAOYSA-N

Application

2,5-Dimethyl-3-pyrroline was used in the synthesis of pyrolyzed M–Nx/C ORR catalyst.

Other Notes

Remainder 2,5-dimethyl-1-pyrroline

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

41.0 °F - closed cup

Flash Point(C)

5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A review on non-precious metal electrocatalysts for PEM fuel cells.
Energy & Environmental Science, 4(9), 3167-3192 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service