149276
5-Nitro-2-furaldehyde diacetate
98%
Synonym(s):
5-Nitrofurfurylidene diacetate
About This Item
Recommended Products
Assay
98%
impurities
<2% 5-nitro-2-furaldehyde
SMILES string
CC(=O)OC(OC(C)=O)c1ccc(o1)[N+]([O-])=O
InChI
1S/C9H9NO7/c1-5(11)15-9(16-6(2)12)7-3-4-8(17-7)10(13)14/h3-4,9H,1-2H3
InChI key
HSXKWKJCZNRMJO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Muta. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service