Skip to Content
Merck
All Photos(2)

Key Documents

146439

Sigma-Aldrich

2-Methyl-4-nitroaniline

97%

Synonym(s):

2-Amino-5-nitrotoluene, 4-Nitro-o-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)NH2
CAS Number:
Molecular Weight:
152.15
Beilstein:
775772
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

130-132 °C (lit.)

functional group

nitro

SMILES string

Cc1cc(ccc1N)[N+]([O-])=O

InChI

1S/C7H8N2O2/c1-5-4-6(9(10)11)2-3-7(5)8/h2-4H,8H2,1H3

InChI key

XTTIQGSLJBWVIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methyl-4-nitroaniline was used in preparing in-plane aligned nanofibers by the electrospinning technique[1]. It was the starting reagent in the synthesis of 3-chloro-5-methylphenyl isocyanate[2]. It was used to deposit polycrystalline thin films on Ag, Cu and Si by conventional and partially ionized beam deposition[3].

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

315.1 °F - closed cup

Flash Point(C)

157.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dmitry V Isakov et al.
ACS nano, 5(1), 73-78 (2010-12-15)
In-plane aligned nanofibers of organic 2-methyl-4-nitroaniline (MNA) were produced by the electrospinning technique using a 1:1 weight ratio with poly(l-lactic acid). The fibers are capable of enormous efficient optical second harmonic generation as strong as pure MNA crystals in powder
3-Fluoro-, 3-chloro-and 3-bromo-5-methylphenylcarbamates of cellulose and amylose as chiral stationary phases for high-performance liquid chromatographic enantioseparation.
Chankvetadze B, et al.
Journal of Chromatography A, 787(1), 67-77 (1997)
Partially ionized beam deposition of 2-methyl-4-nitroaniline thin films.
Nason TC, et al.
Journal of Applied Physics, 70(11), 6766-6773 (1991)
Zhenggong Wang et al.
ChemSusChem, 11(16), 2744-2751 (2018-05-29)
Polyimide-based materials provide attractive chemistries for the development of gas-separation membranes. Modification of inter- and intra-chain interactions is a route to improve the separation performance. In this work, copolyimides with Tröger's base (TB) monomers are designed and synthesized. In particular
Haruna Tahara et al.
The Journal of toxicological sciences, 42(2), 205-221 (2017-03-23)
The ATP assay is a highly sensitive and versatile method for measuring cytotoxicity. However, the correlation between the cell viability results obtained using the ATP assay and those obtained using direct cell counting has not been widely reported. Therefore, to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service