Skip to Content
Merck
All Photos(1)

Key Documents

145297

Sigma-Aldrich

1,3-Dihydroxynaphthalene

99%

Synonym(s):

1,3-Naphthalenediol, Naphthoresorcinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(OH)2
CAS Number:
Molecular Weight:
160.17
Beilstein:
2044002
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

99%

mp

123-125 °C (lit.)

SMILES string

Oc1cc(O)c2ccccc2c1

InChI

1S/C10H8O2/c11-8-5-7-3-1-2-4-9(7)10(12)6-8/h1-6,11-12H

InChI key

XOOMNEFVDUTJPP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Synthon used in the preparation of divinylnaphthalenes and naphthodipyrans.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bulletin of the Chemical Society of Japan, 66, 464-464 (1993)
The Journal of Organic Chemistry, 58, 7388-7388 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service