Skip to Content
Merck
All Photos(1)

Key Documents

144762

Sigma-Aldrich

1,2,5,6,9,10-Hexabromocyclododecane

95%

Synonym(s):

HBCD, HBCDD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18Br6
CAS Number:
Molecular Weight:
641.70
Beilstein:
1911324
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

solubility

acetone: soluble 25 mg/mL, clear, colorless to light yellow
water: slightly soluble

SMILES string

BrC1CCC(Br)C(Br)CCC(Br)C(Br)CCC1Br

InChI

1S/C12H18Br6/c13-7-1-2-8(14)10(16)5-6-12(18)11(17)4-3-9(7)15/h7-12H,1-6H2

InChI key

DEIGXXQKDWULML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2,5,6,9,10-Hexabromocyclododecane(HBCD) is a brominated flame retardant. It is used in the textile industry and polystyrene foam manufacturing. Electrochemical reduction of HBCD at carbon and silver cathodes has been studied using cyclic voltammetry and controlled-potential electrolysis.

Application

1,2,5,6,9,10-Hexabromocyclododecane(HBCD) was used to compare the efficiency of different advanced extraction techniques for the recovery of brominated flame retardants from styrenic polymers. It was used to study the kinetics of the thermal and photolytic segregation of HBCD using HPLC.

Biochem/physiol Actions

1,2,5,6,9,10-Hexabromocyclododecane enhances the diet-induced body weight gain and metabolic dysfunction via disruption of lipid and glucose homeostasis in mice fed normal diet or high-fat diet.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical reduction of 1, 2, 5, 6, 9, 10-hexabromocyclododecane at carbon and silver cathodes in dimethylformamide.
Wagoner ER, et al.
Journal of Electroanalytical Chemistry, 713, 136-142 (2013)
Kinetics of Thermal and Photolytic Segregation of Hexabromocyclododecane in Backcoated Textile Samples.
Ghanem R.
Jordan Journal of Chemistry, 4(2), 171-181 (2009)
Rie Yanagisawa et al.
Environmental health perspectives, 122(3), 277-283 (2014-01-09)
Hexabromocyclododecane (HBCD) is an additive flame retardant used in the textile industry and in polystyrene foam manufacturing. Because of its lipophilicity and persistency, HBCD accumulates in adipose tissue and thus has the potential of causing metabolic disorders through disruption of
Sabine Sévère et al.
SpringerPlus, 4, 27-27 (2015-02-04)
Invasive breast carcinoma is the most common cancer in women as in non-ovariectomised pet dogs, which are already identified as a valuable spontaneous preclinical model for that disease. Geographical and time trends suggest that environmental factors may play an important
Francisco Vilaplana et al.
Journal of chromatography. A, 1196-1197, 139-146 (2008-05-31)
The extraction efficiency of pressurised liquid extraction (PLE), microwave-assisted extraction (MAE), and ultrasonic-assisted extraction (UAE) under different conditions has been compared for the recovery of the most commonly employed brominated flame retardants (BFRs) from styrenic polymeric matrixes. A HPLC-MS/MS method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service