All Photos(1)
About This Item
Linear Formula:
HOC6H4CH=C(CN)CO2H
CAS Number:
Molecular Weight:
189.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
98%
mp
232 °C (dec.) (lit.)
SMILES string
OC(=O)\C(=C\c1cccc(O)c1)C#N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
N Vanderheyden et al.
The Biochemical journal, 346 Pt 1, 53-62 (2000-02-05)
Regulation of intracellular pH (pH(i)) and H(+) efflux were investigated in Trypanosoma brucei bloodstream and procyclic trypomastigotes using the fluorescent dyes 2', 7'-bis-(2-carboxyethyl)-5(6)-carboxyfluorescein (BCECF) acetoxymethyl ester and free BCECF respectively. pH(i) in bloodstream and procyclic trypomastigotes was 7.47+/-0.06 and 7.
E Bassenge et al.
American journal of physiology. Heart and circulatory physiology, 279(5), H2431-H2438 (2000-10-25)
Myocardial ischemia-reperfusion is associated with bursts of reactive oxygen species (ROS) such as superoxide radicals (O(2)(-).). Membrane-associated NADH oxidase (NADHox) activity is a hypothetical source of O(2)(-)., implying the NADH concentration-to-NAD(+) concentration ratio ([NADH]/[NAD(+)]) as a determinant of ROS. To
M Wittner et al.
Pflugers Archiv : European journal of physiology, 402(1), 52-62 (1984-09-01)
Isolated segments of cortical thick ascending limbs (cTAL) of rabbit kidney were perfused in vitro and the equivalent short circuit current (Isc) was measured. In a first series all substrates were removed on either side. Isc fell rapidly to 50
Y H Kang et al.
Molecular and cellular biochemistry, 216(1-2), 37-46 (2001-02-24)
In the hydrogen peroxide (H2O2) apoptosis model of the murine thymocyte, redox reactant and antioxidant pyruvate prevents programmed cell death. We tested the hypothesis that such protection was mediated, at least in part, via pyruvate handling by mitochondrial metabolism. Cultured
R B Hamat et al.
Metabolism: clinical and experimental, 31(6), 548-552 (1982-06-01)
An "in vitro" model of one type of lactic acidosis was produced in rat hemi-diaphragms with inhibitors of pyruvate oxidation. In order to obtain this inhibition in the absence of hypoxia, two actions were sought; inhibiting the mitochondrial pyruvate transporter
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service