Skip to Content
Merck
All Photos(3)

Key Documents

144185

Sigma-Aldrich

2,4-Dichloro-6-methylpyrimidine

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€88.20
100 G
€139.00

€88.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€88.20
100 G
€139.00

About This Item

Empirical Formula (Hill Notation):
C5H4Cl2N2
CAS Number:
Molecular Weight:
163.00
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€88.20


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

bp

219 °C (lit.)

mp

44-47 °C (lit.)

functional group

chloro

SMILES string

Cc1cc(Cl)nc(Cl)n1

InChI

1S/C5H4Cl2N2/c1-3-2-4(6)9-5(7)8-3/h2H,1H3

InChI key

BTLKROSJMNFSQZ-UHFFFAOYSA-N

General description

2,4-Dichloro-6-methylpyrimidine undergoes double cross-coupling reaction with 2-(tributylstannyl)pyridine, followed by aldol condensation to yield 4-arylvinyl-2,6-di(pyridin-2-yl)pyrimidines[1]. It reacts with 1H,1H,2H,2H-perfluorodecanethiol during fluorous synthesis of disubstituted pyrimidines[2].

Application

2,4-Dichloro-6-methylpyrimidine was used in the synthesis of (2-chloro-6-methyl-pyrimidin-4-yl)-(2,3-dihydro-benzothiazol-6-yl)-amine[3].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caroline Hadad et al.
The Journal of organic chemistry, 76(10), 3837-3845 (2011-04-06)
A series of 4-arylvinyl-2,6-di(pyridin-2-yl)pyrimidines have been efficiently prepared by a double cross-coupling reaction between 2,4-dichloro-6-methylpyrimidine and 2-(tributylstannyl)pyridine, followed by aldol condensation with the appropriate aromatic aldehyde substituted with electron-donating, electron-withdrawing, dendritic, or water-soluble groups. The effect of different protic and
Eliud Hernández et al.
Puerto Rico health sciences journal, 29(4), 348-356 (2011-01-26)
Rho family GTPases are molecular switches that control signaling pathways regulating a myriad of cellular functions. Rac1, a Rho family member, plays a critical role in several aspects of tumorigenesis, cancer progression, invasion, and metastasis. Rac proteins are not mutated
Wei Zhang
Organic letters, 5(7), 1011-1013 (2003-03-28)
[reaction: see text] The fluorous synthesis of disubstituted pyrimidines is carried out by attaching 2,4-dichloro-6-methylpyrimidine with 1H,1H,2H,2H-perfluorodecanethiol. The tagged substrate is substituted with 3-(trifluoromethyl)pyrazole followed by thioether oxidation and tag displacement with amines or thiols. The fluorous chain serves as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service