Skip to Content
Merck
All Photos(1)

Key Documents

143081

Sigma-Aldrich

Phenyl isocyanide dichloride

98%

Synonym(s):

N-(Dichloromethylene)aniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N=CCl2
CAS Number:
Molecular Weight:
174.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

liquid

refractive index

n20/D 1.571 (lit.)

bp

103-106 °C/30 mmHg (lit.)

density

1.265 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

Cl\C(Cl)=N\c1ccccc1

InChI

1S/C7H5Cl2N/c8-7(9)10-6-4-2-1-3-5-6/h1-5H

InChI key

TTWWZVGVBRPHLE-UHFFFAOYSA-N

Application

Phenyl isocyanide dichloride was used in the synthesis of 5-halo-1-phenyltetrazoles[1].

Biochem/physiol Actions

Phenyl isocyanide dichloride exhibits spectral interactions with cytochrome P-450 from mouse hepatic microsomes[2].

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

5-Halo-1-phenyltetrazoles.
Collibee WL, et al.
The Journal of Organic Chemistry, 60(2), 468-469 (1995)
G A Beumel et al.
General pharmacology, 16(3), 193-197 (1985-01-01)
The binding of isocyanides and the metabolites of piperonyl butoxide (PBO) to reduced cytochrome P-450 in intact microsomes gives rise to the type III optical difference spectrum which is characterized by two pH dependent peaks in the Soret region. Each
G A Beumel et al.
General pharmacology, 16(3), 189-192 (1985-01-01)
The spectral interactions of a number of isocyanides with cytochrome P-450 were investigated. An interaction between the hydrophobic nature of the side chain and the spectral interaction was apparent. The concentration of the isocyanide affected the stability of the complex.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service