Skip to Content
Merck
All Photos(2)

Key Documents

141879

Sigma-Aldrich

(−)-Lobeline hydrochloride

98%

Synonym(s):

α-Lobeline hydrochloride, L-Lobeline hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€263.00

€263.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€263.00

About This Item

Empirical Formula (Hill Notation):
C22H27NO2 · HCl
CAS Number:
Molecular Weight:
373.92
Beilstein:
3920196
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€263.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

optical activity

[α]20/D −56.8°, c = 2 in H2O

mp

183-185 °C (dec.) (lit.)

functional group

hydroxyl
ketone
phenyl

SMILES string

Cl[H].CN1[C@@H](CCC[C@@H]1CC(=O)c2ccccc2)C[C@H](O)c3ccccc3

InChI

1S/C22H27NO2.ClH/c1-23-19(15-21(24)17-9-4-2-5-10-17)13-8-14-20(23)16-22(25)18-11-6-3-7-12-18;/h2-7,9-12,19-21,24H,8,13-16H2,1H3;1H/t19-,20+,21-;/m0./s1

InChI key

MKMYPTLXLWOUSO-NFQNBQCWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeong-Ho Park et al.
FEBS letters, 590(4), 493-500 (2016-01-24)
The intestine is involved in digestion and absorption, as well as the regulation of metabolism upon sensation of the internal intestinal environment. Enteroendocrine cells are thought to mediate these internal intestinal chemosensory functions. Using the CaLexA (calcium-dependent nuclear import of
Hany Km Dweck et al.
eLife, 10 (2021-02-23)
Although most Drosophila species lay eggs in overripe fruit, the agricultural pest Drosophila suzukii lays eggs in ripe fruit. We found that changes in bitter taste perception have accompanied this adaptation. We show that bitter-sensing mutants of Drosophila melanogaster undergo
Jaekyun Choi et al.
eNeuro, 7(2) (2020-03-30)
Feeding, a critical behavior for survival, consists of a complex series of behavioral steps. In Drosophila larvae, the initial steps of feeding are food choice, during which the quality of a potential food source is judged, and ingestion, during which
Michelle Bui et al.
BMC neuroscience, 20(1), 27-27 (2019-06-19)
The mosquito Aedes aegypti has a wide variety of sensory pathways that have supported its success as a species as well as a highly competent vector of numerous debilitating infectious pathogens. Investigations into mosquito sensory systems and their effects on
Hany K M Dweck et al.
Current biology : CB, 30(1), 17-30 (2019-12-17)
Taste systems detect a vast diversity of toxins, which are perceived as bitter. When a species adapts to a new environment, its taste system must adapt to detect new death threats. We deleted each of six commonly expressed bitter gustatory receptors (Grs)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service