Skip to Content
Merck
All Photos(1)

Documents

141682

Sigma-Aldrich

2-Thiopheneacetonitrile

97%

Synonym(s):

2-Thienylacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NS
CAS Number:
Molecular Weight:
123.18
Beilstein:
106960
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.542 (lit.)

bp

115-120 °C/22 mmHg (lit.)
235-238 °C (lit.)

density

1.157 g/mL at 25 °C (lit.)

SMILES string

N#CCc1cccs1

InChI

1S/C6H5NS/c7-4-3-6-2-1-5-8-6/h1-2,5H,3H2

InChI key

CLSHQIDDCJTHAJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Thiopheneacetonitrile was used as reagent during the synthesis of monodisperse Au/Au@polythiophene core/shell nanospheres.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hye-Seon Shin et al.
ACS applied materials & interfaces, 4(11), 6324-6331 (2012-10-31)
Monodisperse Au/Au@polythiophene core/shell nanospheres were facilely prepared through the reduction of gold precursor, AuCl₄⁻, by 2-thiopheneacetonitrile in an aqueous solution. Concomitantly, 2-thiopheneacetonitrile polymerized during this redox process. As a result, Au nanoparticle was encapsulated by conductive polymer shell to afford

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service