Skip to Content
Merck
All Photos(1)

Key Documents

141240

Sigma-Aldrich

2-Chloro-6-fluorobenzaldehyde

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(F)CHO
CAS Number:
Molecular Weight:
158.56
Beilstein:
2245530
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

form

solid

mp

32-35 °C (lit.)

functional group

aldehyde
chloro
fluoro

SMILES string

[H]C(=O)c1c(F)cccc1Cl

InChI

1S/C7H4ClFO/c8-6-2-1-3-7(9)5(6)4-10/h1-4H

InChI key

OACPOWYLLGHGCR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Chloro-6-fluorobenzaldehyde undergoes piperidine catalyzed Knoevenagel condensation reaction with methyl cyanoacetate to yield methyl 2-cyano-3-dihalophenyl-2-propenoate[1].

Application

2-Chloro-6-fluorobenzaldehyde was used in the synthesis of novel copolymers of methyl 2-cyano-3-dihalophenyl-2-propenoates and styrene[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel copolymers of trisubstituted ethylenes with styrene-7. Methyl 2-cyano-3-dihalophenyl-2-propenoates.
Kharas GB, et al.
Polym. Bull., 45(4-5), 351-357 (2000)

Articles

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service